Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SCO1 Synthetic Peptide

Catalog No. PEP0465
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0465 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0465 Supplier Invitrogen™ Supplier No. PEP0465
Add to Cart
Add to Cart

Synthetic Peptide

This peptide corresponds to 14 amino acids near the center of human SCO1 PEP-0465 can be used as a blocking peptide with polyclonal antibody PA5-20345.

Synthesis of cytochrome c oxidase 1 was initially identified in yeast as one of two cytochrome c oxidase (COX) assembly proteins that enable the assembly of cytochrome c holoenzyme, a complex that catalyzes the transfer of reducing equivalents from cytochrome c to molecular oxygen and pumps protons across the inner mitochondrial membrane. Like their yeast homologs, the function of both SCO1 and SCO2 are dependent on copper ion binding. Mutations in either gene can lead to cytochrome c oxidase respiratory chain defects, with a missense mutation in human SCO1 (P174L) associated with a fatal neonatal hepatopathy when the second allele is also non-functional, suggesting the pathology is due to loss of function. It has been suggested that this mutation alters the SCO1 affinity for the copper (I) ion, thus impairing the efficiency of copper transfer to the cytochrome c oxidase.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 6341
Species Human
Purification Method purified
Accession Number O75880
Common Name SCO1
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Alias 2610001C07Rik; D11Bwg1310e; Protein SCO1 homolog, mitochondrial; RGD1559538; SCO cytochrome c oxidase assembly protein 1; SCO cytochrome oxidase deficient homolog 1; SCO cytochrome oxidase deficient homolog 1 (yeast); SCO1; SCO1 cytochrome c oxidase assembly protein; SCO1, cytochrome c oxidase assembly protein; SCOD1; synthesis of cytochrome C oxidase 1
Gene Symbol SCO1
Lyophilized No
Preservative Sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.