Learn More
Sino Biological Human SDC1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 11429H02H100
Encompass_Preferred
A DNA sequence encoding the Human SDC1 (NP_002988.3) (Met1-Glu251) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-1702