Learn More
Sino Biological Recombinant Human Syndecan-1/CD138 Protein (ECD, His & AVI Tag), Biotinylated
Supplier:  Sino Biological 11429H49HB20
Encompass_Preferred
A DNA sequence encoding the Human SDC1 (NP_002988.3) (Met1-Glu251) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-8446