Learn More
Sino Biological Anti-Syndecan-2/CD362 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 207417T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Syndecan-2 / SDC2 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3864