Learn More
Sino Biological Anti-Syndecan-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12158T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Syndecan-3 / SDC3 (rh Syndecan-3 / SDC3; Catalog#12158-H08H; ABM82826.1; Met1-Lys311). Syndecan-3 / SDC3 specific IgG was purified by Human Syndecan-3 / SDC3 affinity chromatography.
Catalog No. 50-255-4452