Learn More
ACROBiosystems ACROBiosystems Human SDF-1 / CXCL12 Protein, His Tag
Catalog No. 502106806
Encompass_Preferred
Recombinant Protein;50UG;Human SDF-1, His Tag (SD1-H5143) is expressed from E.coli cells. It contains AA Lys 22 - Lys 89 (Accession # P48061-2).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 10.0 kDa. The protein migrates as 13 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-210-6806 Supplier ACROBiosystems Supplier No. SD1H514350UG
Recombinant Protein;50UG;Human SDF-1, His Tag (SD1-H5143) is expressed from E.coli cells. It contains AA Lys 22 - Lys 89 (Accession # P48061-2).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 10.0 kDa. The protein migrates as 13 kDa under reducing (R) condition (SDS-PAGE).