Learn More
Biomatik Corporation Human Secernin 1 (SCRN1) ELISA Kit
Supplier:  Biomatik Corporation EKU12517
Encompass_Preferred
Human Secernin 1 (SCRN1) ELISA Kit for detecting SCRN1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q12765. Gene ID: 9805. Target Synonyms: SES1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7953