Learn More
Biomatik Corporation Human Secretagogin (SCGN) ELISA Kit
Supplier:  Biomatik Corporation EKU07209
Encompass_Preferred
Human Secretagogin (SCGN) ELISA Kit for detecting SCGN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O76038. Gene ID: 10590. Target Synonyms: SECRET; SEGN; CALBL; Calbindin-Like; Secretagogin,EF-Hand Calcium Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8771