Learn More
American Research Products Inc Human secretory component SC
Catalog No. 5000328786
Encompass
Human secretory component SC ELISA Kit from CUSABIO Detection Range 0 78 ng/ml-50 ng/ml Sensitivity 0 195 ng/ml Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28786 Supplier American Research Products Inc Supplier No. CSBE09016H
Human secretory component SC ELISA Kit from CUSABIO Detection Range 0 78 ng/ml-50 ng/ml Sensitivity 0 195 ng/ml Sample serum plasma cell culture supernates tissue homogenates Method Sandwich