Learn More
ACROBiosystems ACROBiosystems Human Secretory Component, Twin Strep Tag
Catalog No. 502106786
Encompass_Preferred
Recombinant Protein;25UG;Human Secretory Component, Twin Strep Tag (SCT-H5280) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Arg 603 (Accession # P01833-1).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 67.6 kDa. The protein migrates as 80-100 kDa, NR:80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6786 Supplier ACROBiosystems Supplier No. SCTH528025UG
Recombinant Protein;25UG;Human Secretory Component, Twin Strep Tag (SCT-H5280) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Arg 603 (Accession # P01833-1).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 67.6 kDa. The protein migrates as 80-100 kDa, NR:80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.