Learn More
Sino Biological Recombinant Anti-E-Selectin/CD62E Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10335R04020
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD62E / E-selectin (rh CD62E; Catalog#10335-H08H; NP_000441.2; Met 1-Pro 556).
Catalog No. 50-240-9053