Learn More
American Research Products Inc Human selenium binding protein
Catalog No. 5000329739
Encompass
Human selenium binding protein 1 (SELENBP1) ELISA Kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29739 Supplier American Research Products Inc Supplier No. CSBE13947H
Human selenium binding protein 1 (SELENBP1) ELISA Kit from CUSABIO Detection Range 125 pg/mL-8000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich