Learn More
Biomatik Corporation Human Selenoprotein W1 (SEPW1) ELISA Kit
Supplier:  Biomatik Corporation EKU12231
Encompass_Preferred
Human Selenoprotein W1 (SEPW1) ELISA Kit for detecting SEPW1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P63302. Gene ID: 6415. Target Synonyms: SEP-W1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7998