Learn More
Sino Biological Recombinant Anti-L-selectin/CD62L Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 11838R018H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD62L / L-Selectin / SELL (rh CD62L / L-Selectin / SELL; Catalog#11838-H08H; P14151-1; Met 1-Asn 332), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-8984