Learn More
Sino Biological Recombinant Anti-L-selectin/CD62L Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 11838R371F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SELL / CD62L (rh SELL / CD62L; Catalog#11838-H08H; P14151-1; Met1-Asn332) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-9117