Learn More
Sino Biological Anti-L-selectin/CD62L Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 11838MM01A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD62L / L-Selectin (rh CD62L / L-Selectin; Catalog#11838-H08H; P14151-1; Met1-Asn332) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-1432