Learn More
Sino Biological Recombinant Anti-L-selectin/CD62L Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 11838R220P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SELL / CD62L (rh SELL / CD62L; Catalog#11838-H08H; P14151-1; Met1-Asn332), and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4525