Learn More
Sino Biological Anti-L-selectin/CD62L Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11838T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD62L/L-Selectin (rh CD62L/L-Selectin; Catalog#11838-H08H; P14151-1; Met1-Asn332). CD62L/L-Selectin specific IgG was purified by Human CD62L/L-Selectin affinity chromatography.
Catalog No. 50-240-9573