Learn More
Sino Biological Human SELL PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11838RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD62L / L-Selectin / SELL (rh CD62L / L-Selectin / SELL; Catalog#11838-H08H; P14151-1; Met 1-Asn 332). CD62L / L-Selectin / SELL specific IgG was purified by Human CD62L / L-Selectin / SELL affinity chromatography.
Catalog No. 50-237-2774