Learn More
Sino Biological Anti-P-Selectin/CD62P Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13025RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SELP / P Selectin (rh SELP; Catalog#13025-H08H; AAN06828.1; Met1-Ala771). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0024