Learn More
Sino Biological Recombinant Anti-PSGL-1/CD162 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 13863R031F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PSGL-1 / CD162 (rh PSGL-1 / CD162; Catalog#13863-H08H; Q14242-1; Met1-Gly295) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8963