Learn More
Sino Biological Recombinant Anti-PSGL-1/CD162 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 13863R031A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PSGL-1 / CD162 / SELPLG (rh PSGL-1 / CD162 / SELPLG; Catalog#13863-H08H; Q14242-1; Met1-Gly295) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-9244