Learn More
Sino Biological Anti-PSGL-1/CD162 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 13863MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PSGL-1 / CD162 / SELPLG (rh PSGL-1 / CD162 / SELPLG; Catalog#13863-H08H; Q14242-1; Met1-Gly295). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1965