Learn More
Sino Biological Recombinant Anti-PSGL-1/CD162 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 13863R061100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PSGL-1 / CD162 / SELPLG (rh PSGL-1 / CD162 / SELPLG; Catalog#13863-H08H; Q14242-1; Met1-Gly295).
Catalog No. 50-240-8088