Learn More
Sino Biological Anti-PSGL-1/CD162 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 13863MM07P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PSGL-1 / CD162 (rh PSGL-1 / CD162; Catalog#13863-H08H; Q14242-1; Met1-Gly295) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-2402