Learn More
Sino Biological Human SELPLG MAb 100ul (Sino Biological)
Supplier:  Sino Biological 13863MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PSGL-1 / CD162 / SELPLG (rh PSGL-1 / CD162 / SELPLG; Catalog#13863-H08H; Q14242-1; Met1-Gly295). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2414