Learn More
Sino Biological Anti-Semaphorin 4D Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11825MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Semaphorin 4D / SEMA4D / CD100 (rh Semaphorin 4D / SEMA4D / CD100; Catalog#11825-H08H; AAH54500.1; Met1-Arg734). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4538