Learn More
Sino Biological Recombinant Anti-Semaphorin 4D Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11825R301100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Semaphorin 4D / SEMA4D / CD100 (rh Semaphorin 4D / SEMA4D / CD100; Catalog#11825-H08H; AAH54500.1; Met1-Arg734).
Catalog No. 50-255-4534