Learn More
Sino Biological Recombinant Human Semaphorin 4D Protein (ECD, hFc Tag)
Supplier:  Sino Biological 11825H02H200
Encompass_Preferred
A DNA sequence encoding the human SEMA4D (AAH54500.1) (Met1-Arg734) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4540