Learn More
Biomatik Corporation Human Semaphorin 3D (SEMA3D) ELISA Kit
Supplier:  Biomatik Corporation EKU12087
Encompass_Preferred
Human Semaphorin 3D (SEMA3D) ELISA Kit for detecting SEMA3D in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.34ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95025. Gene ID: 223117. Target Synonyms: Coll-2; Sema-Z2; Sema Domain,Immunoglobulin Domain (Ig) ,Transmembrane Domain (TM) and Short Cytoplasmic Domain 3D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5578