Learn More
Invitrogen™ Human Semaphorin 6B ELISA Kit
Description
Human Semaphorin 6B quantitates human Semaphorin 6B in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Semaphorin 6B.
Specifications
Specifications
| Accession Number | Q9H3T3 |
| Assay Range | 0.6-150 ng/mL |
| Assay Sensitivity | 0.6 ng/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | Sema; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B; sema N; Sema VIB; sema Z; SEMA6B; SEMAN; semaphorin 6B; semaphorin VIB; semaphorin Z; semaphorin-6B; semaphorin-6Ba; semaphorin-N; Semaphorin-Z; SEMA-VIB; semaZ; SEM-SEMA-Y; UNQ1907/PRO4353; VIbSema; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B; sema N; Sema VIB; sema Z; SEMA6B; SEMAN; semaphorin 6B; semaphorin VIB; semaphorin Z; semaphorin-6B; semaphorin-6Ba; semaphorin-N; Semaphorin-Z; SEMA-VIB; semaZ; SEM-SEMA-Y; UNQ1907/PRO4353; VIb; |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.