Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Semaphorin 6D ELISA Kit

Catalog No. EH409RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH409RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH409RBX10 Supplier Invitrogen™ Supplier No. EH409RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human Semaphorin 6D quantitates human Semaphorin 6D in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Semaphorin 6D.

Semaphorins are a large family, including both secreted and membrane associated proteins, many of which have been implicated as inhibitors or chemorepellents in axon pathfinding, fasciculation and branching, and target selection. All semaphorins possess a semaphorin (Sema) domain and a PSI domain (found in plexins, semaphorins and integrins) in the N-terminal extracellular portion. Additional sequence motifs C-terminal to the semaphoring domain allow classification into distinct subfamilies. Results demonstrate that transmembrane semaphorins, like the secreted ones, can act as repulsive axon guidance cues. SEMA6D is a class 6 vertebrate transmembrane semaphorin that demonstrates alternative splicing. Six transcript variants have been identified and expression of the distinct encoded isoforms is thought to be regulated in a tissue- and development-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NFY4
Assay Range 1.64-400 ng/mL
Assay Sensitivity 1.64 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 1110067B02Rik; AA409156; D330011G23; FLJ11598; hypothetical protein LOC406272; KIAA1479; KIAA1479FLJ11598; mKIAA1479; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D; Sema6d; sema6d protein; semaphorin 6D; semaphorin-6D; wu:fi06a12; wu:fi76a10; zgc:561701110067B02Rik; AA409156; D330011G23; FLJ11598; hypothetical protein LOC406272; KIAA1479; KIAA1479FLJ11598; mKIAA1479; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D; Sema6d; sema6d protein; semaphorin 6D; semaphorin-6D; wu:fi06a12; wu:fi76a10; zgc:56170;
Gene ID (Entrez) 80031
Gene Symbol SEMA6D
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target SEMA6D
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.