Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Semaphorin-7A ELISA Kit

Catalog No. EH410RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH410RB 96 Tests
EH410RBX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH410RB Supplier Invitrogen™ Supplier No. EH410RB
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human Semaphorin-7A quantitates human Semaphorin-7A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Semaphorin-7A.

CD108 (Sema7A) is a GPI-anchored semaphorin family member, which enhances central and peripheral axonal growth and is required for proper axon track formation during ebryogenesis. CD108 also regulates osteoclast differentiation and pre-osteoblastic cell migration, and in immune system affects cell proliferation, chemotaxis and cytokine release. On erythrocytes CD108 defines the JMH (John-Milton-Hagen) human blood group. CD108 signalizes through its receptors - plexin C1 and beta1 integrins.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75326
Assay Range 0.16-40 ng/mL
Assay Sensitivity 0.16 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 2900057C09Rik; CD108; CDw108; H-Sema K1; H-SEMA-K1; H-Sema-L; JMH; JMH blood group antigen; John Milton Hagen blood group H-Sema K1; john-Milton-Hargen human blood group Ag; MGC126692; MGC126696; M-Sema-L; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A (JMH blood group); sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A; sema K1; Sema L; SEMA7A; SEMAK1; SEMAL; semaphorin 7A (John Milton Hagen blood group); semaphorin 7A, GPI membrane anchor; semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group); Semaphorin K1; semaphorin-7A; Semaphorin-K1; semaphorin-L; Semk12900057C09Rik; CD108; CDw108; H-Sema K1; H-SEMA-K1; H-Sema-L; JMH; JMH blood group antigen; John Milton Hagen blood group H-Sema K1; john-Milton-Hargen human blood group Ag; MGC126692; MGC126696; M-Sema-L; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A (JMH blood group); sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A; sema K1; Sema L; SEMA7A; SEMAK1; SEMAL; semaphorin 7A (John Milton Hagen blood group); semaphorin 7A, GPI membrane anchor; semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group); Semaphorin K1; semaphorin-7A; Semaphorin-K1; semaphorin-L; Semk1;
Gene ID (Entrez) 8482
Gene Symbol SEMA7A
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target SEMA7A
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.