Learn More
Biomatik Corporation Human Semenogelin II (SEMG2) ELISA Kit
Supplier:  Biomatik Corporation EKU07272
Encompass_Preferred
Human Semenogelin II (SEMG2) ELISA Kit for detecting SEMG2 in tissue homogenates, seminal plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q02383. Gene ID: 6407. Target Synonyms: SGII. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8767