Learn More
Biomatik Corporation Human Septin 5 (SEPT5) ELISA Kit
Supplier:  Biomatik Corporation EKU07277
Encompass_Preferred
Human Septin 5 (SEPT5) ELISA Kit for detecting SEPT5 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99719. Gene ID: 5413. Target Synonyms: CDCREL; CDCREL-1; CDCREL1; H5; PNUTL1; Peanut-Like 1; Cell division control-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7666