Learn More
Biomatik Corporation Human Septin 6 (SEPT6) ELISA Kit
Supplier:  Biomatik Corporation EKU07278
Encompass_Preferred
Human Septin 6 (SEPT6) ELISA Kit for detecting SEPT6 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14141. Gene ID: 23157. Target Synonyms: 45171. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8766