Learn More
American Research Products Inc Human Serine threonine protein
Catalog No. 5000327084
Encompass
Human Serine/threonine-protein kinase LMTK3 (LMTK3) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 7 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27084 Supplier American Research Products Inc Supplier No. CSBEL013017HU
Human Serine/threonine-protein kinase LMTK3 (LMTK3) ELISA kit from CUSABIO Detection Range 18 75 pg/mL-1200 pg/mL Sensitivity 4 7 pg/mL Sample serum plasma tissue homogenates Method Sandwich