Learn More
Bio Rad Laboratories BUF070B, Human Seroblock, 200 Tests
Catalog No. NC2952698
Encompass
Human Fc receptors are expressed on a variety of immune cell types including monocytes, macrophages, B cells, granulocytes and dendritic cells. Cells that express Fc receptors can give false positive immunofluorescent staining due to the Fc receptors binding of Ig. Human SeroBlock is designed to prevent such non-specific staining without interfering with appropriate target staining. Human SeroBlock is compatible with use of anti-human antibodies targeting Fc receptors in flow cytometry.
Catalog No. NC2952698 Supplier Bio Rad Laboratories Supplier No. BUF070B
Human Fc receptors are expressed on a variety of immune cell types including monocytes, macrophages, B cells, granulocytes and dendritic cells. Cells that express Fc receptors can give false positive immunofluorescent staining due to the Fc receptors binding of Ig. Human SeroBlock is designed to prevent such non-specific staining without interfering with appropriate target staining. Human SeroBlock is compatible with use of anti-human antibodies targeting Fc receptors in flow cytometry.