Learn More
ACROBiosystems ACROBiosystems Human Serpin A3 Protein, His Tag
Catalog No. 502106766
Encompass_Preferred
Recombinant Protein;50UG;Human Serpin A3, His Tag (SE3-H5221) is expressed from human 293 cells (HEK293). It contains AA His 24 - Ala 423 (Accession # AAH03559).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 46.6 kDa. The protein migrates as 50-63 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6766 Supplier ACROBiosystems Supplier No. SE3H522150UG
Recombinant Protein;50UG;Human Serpin A3, His Tag (SE3-H5221) is expressed from human 293 cells (HEK293). It contains AA His 24 - Ala 423 (Accession # AAH03559).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 46.6 kDa. The protein migrates as 50-63 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.