Learn More
Biomatik Corporation Human Serpin B3 (SERPINB3) CLIA Kit
Supplier:  Biomatik Corporation EKU08939
Encompass_Preferred
Human Serpin B3 (SERPINB3) CLIA Kit for detecting SERPINB3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9BYH9. Target Synonyms: SCCA1; T4-A; Serpin Peptidase Inhibitor,Clade B (ovalbumin) ,Member 3; Serpin B3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5424