Learn More
Raybiotech Inc Human SERPINF1 (PEDF) ELISA Kit
Supplier:  Raybiotech Inc ELHSERPINF11
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.6 ug/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit detects human Serpin F1. Approximately 40% cross-reactivity with recombinant mouse Serpin F1 is observed in this antibody pair.
Catalog No. 50-194-9717