Learn More
ACROBiosystems Human Serpin G1 / C1 inhibitor Protein, His Tag
Catalog No. 502331375
Encompass_Preferred
Human Serpin G1 Protein, His Tag (SER-H52H5) is expressed from human 293 cells (HEK293). It contains AA Asn 23 - Ala 500 (Accession # P05155-1).
Catalog No. 50-233-1375 Supplier ACROBiosystems Supplier No. SERH52H5200UG
Human Serpin G1 Protein, His Tag (SER-H52H5) is expressed from human 293 cells (HEK293). It contains AA Asn 23 - Ala 500 (Accession # P05155-1).