Learn More
Biomatik Corporation Human Serpin Peptidase Inhibitor Clade B Member 10 (SERPINB10) ELISA Kit
Supplier:  Biomatik Corporation EKU11422
Encompass_Preferred
Human Serpin Peptidase Inhibitor Clade B Member 10 (SERPINB10) ELISA Kit for detecting SERPINB10 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8,000pg/ml. Sensitivity: 54pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48595. Gene ID: 5273. Target Synonyms: Bomapin; Peptidase inhibitor 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8145