Learn More
Sino Biological Recombinant Human Serpin A1/Alpha-1-antitrypsin Protein (His Tag), HPLC-verified 1 mg
Supplier:  Sino Biological 10306H08H1
Encompass_Preferred
A DNA sequence encoding the human SerpinA1 (NP_000286.3) pre-protein (Met 1-Lys 418) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-224-6653