Learn More
Sino Biological Human SerpinA1 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10306R00520
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SerpinA1 (rh SerpinA1; Catalog#10306-H08H; NP_000286.3; Met1-Lys418).
Catalog No. 50-237-2215