Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SERPINA1 native Protein from human plasma

Catalog No. 89961497 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-497 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-961-497 Supplier Abnova™ Supplier No. P4948

Used for SDS-PAGE

The protein encoded by this gene is secreted and is a serine protease inhibitor whose targets include elastase, plasmin, thrombin, trypsin, chymotrypsin, and plasminogen activator. Defects in this gene can cause emphysema or liver disease. Several transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Specifications

For Use With (Application) SDS-PAGE
Formulation Lyophilized from 30mM sodium phosphate, 300mM NaCl, pH 6.5
Gene ID (Entrez) 5265
Molecular Weight (g/mol) 52kDa
Name SERPINA1 (Human) Native Protein
Preparation Method Native protein purified from human plasma
Quantity 1 mg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias A1A/A1AT/AAT/MGC23330/MGC9222/PI/PI1/PRO2275
Common Name SERPINA1
Gene Symbol SERPINA1
Species Human
Protein Tag None
Expression System Native protein purified from human plasma
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.