Learn More
Sino Biological Recombinant Human Serpina12 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11822H08H1
Encompass_Preferred
A DNA sequence encoding the human SERPINA12 (NP_776249.1) (Met 1-Lys 414) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4541