Learn More
Sino Biological Recombinant Human SerpinA3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10307H08H100
Encompass_Preferred
A DNA sequence encoding the human SERPINA3 (NP_001076.2) (Met 1-Ala 423) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-224-7273