Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SERPINA3 native Protein from human plasma

Catalog No. 89961506 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-961-506 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-961-506 Supplier Abnova™ Supplier No. P4947

Used for SDS-PAGE

The protein encoded by this gene is a plasma protease inhibitor and member of the serine protease inhibitor class. Polymorphisms in this protein appear to be tissue specific and influence protease targeting. Variations in this protein's sequence have been implicated in Alzheimer's disease, and deficiency of this protein has been associated with liver disease. Mutations have been identified in patients with Parkinson disease and chronic obstructive pulmonary disease. [provided by RefSeq]

Specifications

For Use With (Application) SDS-PAGE
Formulation Lyophilized from 20mM Tris-HCl, 150mM NaCl, pH 7.4
Gene ID (Entrez) 12
Molecular Weight (g/mol) 64kDa
Name SERPINA3 (Human) Native Protein
Preparation Method Native protein purified from human plasma
Quantity 100 μg
Storage Requirements Store at -20°C on dry atmosphere. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AACT/ACT/GIG24/GIG25/MGC88254
Common Name SERPINA3
Gene Symbol SERPINA3
Species Human
Protein Tag None
Expression System Native protein purified from human plasma
Form Lyophilized
Purity or Quality Grade ≥95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.