Learn More
Cusabio Technology LLC HUMAN SERPINA6 ELISA KIT
Catalog No. 5000050836
Encompass_Preferred
The Human Corticosteroid Binding Globulin (CBG) ELISA Kit is designed to quantitatively measure SERPINA6 (Corticosteroid Binding Globulin) levels in serum plasma and tissue homogenates using a competitive assay format This kit detects SERPINA6 with a sensitivity of 20 pmol/mL and offers a broad detection range from 20 pmol/mL to 8000 pmol/mL making it suitable for both low and high concentration samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours with absorbance measurements taken at 450 nm Its reliable performance and quantitative accuracy support applications in clinical research and biomarker studies related to steroid hormone transport and regulation
Catalog No. 50-000-50836 Supplier Cusabio Technology LLC Supplier No. CSBE14214H
The Human Corticosteroid Binding Globulin (CBG) ELISA Kit is designed to quantitatively measure SERPINA6 (Corticosteroid Binding Globulin) levels in serum plasma and tissue homogenates using a competitive assay format This kit detects SERPINA6 with a sensitivity of 20 pmol/mL and offers a broad detection range from 20 pmol/mL to 8000 pmol/mL making it suitable for both low and high concentration samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours with absorbance measurements taken at 450 nm Its reliable performance and quantitative accuracy support applications in clinical research and biomarker studies related to steroid hormone transport and regulation